ContactUsLinkComponent
Glass therm., industrial, V-shape, DIN 16181, 0+100:2°C, type B, built-in length 63mm, special liquid filled, upper part 110mm, smal l straight pattern, thread G 1/2 A 1 * 1 items
Catalog # AMART123010-FL
Supplier:  Amarell
Glass therm., industrial, V-shape, DIN 16181, 0+100:2°C, type B, built-in length 63mm, special liquid filled, upper part 110mm, smal l straight pattern, thread G 1/2 A 1 * 1 items
Catalog # AMART123010-FL
Supplier:  Amarell
Frequently Bought Together