ContactUsLinkComponent
Straight stem therm., fine subdivision, 0+100:0,5°C, milk glass scale, cap. prismatic, yellow backed, red special liquid, upper part 250x16-17mm, lower part 300x8-8,5mm, pin-top finish, suitable for government verification 1 * 1 items
Catalog # AMART104166-FL
Supplier:  Amarell
Straight stem therm., fine subdivision, 0+100:0,5°C, milk glass scale, cap. prismatic, yellow backed, red special liquid, upper part 250x16-17mm, lower part 300x8-8,5mm, pin-top finish, suitable for government verification 1 * 1 items
Catalog # AMART104166-FL
Supplier:  Amarell
Frequently Bought Together