ContactUsLinkComponent
Gallium-Quartz-Glass therm., 0+1050:5°C, upper part c. 300x13-15mm, lower part 200x8-9mm, milk-glass scale, cap. colourless, partial immersion is by 5-10cm shorter than the stem, cork-plaster-top finish, with metal cap 1 * 1 items
Catalog # AMARQ140202-GA
Supplier:  Amarell
Gallium-Quartz-Glass therm., 0+1050:5°C, upper part c. 300x13-15mm, lower part 200x8-9mm, milk-glass scale, cap. colourless, partial immersion is by 5-10cm shorter than the stem, cork-plaster-top finish, with metal cap 1 * 1 items
Catalog # AMARQ140202-GA
Supplier:  Amarell
Frequently Bought Together