ContactUsLinkComponent
Therm. to set Amarell Universal, +350+402:0,1°C, 400x9mm, enclosed scale, cap.lens front, colourless, gallium filled, total immersio n, richter-top finish, government calibrated with Calibration Certificate with 3 test points 1 * 1 items
Catalog # AMARN71020-GA/CC
Supplier:  Amarell
Therm. to set Amarell Universal, +350+402:0,1°C, 400x9mm, enclosed scale, cap.lens front, colourless, gallium filled, total immersio n, richter-top finish, government calibrated with Calibration Certificate with 3 test points 1 * 1 items
Catalog # AMARN71020-GA/CC
Supplier:  Amarell
Frequently Bought Together