ContactUsLinkComponent
Therm. to set Amarell Universal, +97+152:0,1C, 400x9mm, enclosed scale, capillary tube specially coated inside, with non-wetting b lue special liquid, total immersion, richter-top finish, null
Catalog # AMARN71010-CB/WC
Supplier:  Amarell
Therm. to set Amarell Universal, +97+152:0,1C, 400x9mm, enclosed scale, capillary tube specially coated inside, with non-wetting b lue special liquid, total immersion, richter-top finish, null
Catalog # AMARN71010-CB/WC
Supplier:  Amarell
Frequently Bought Together