ContactUsLinkComponent
Therm. to set Amarell Universal, +48+102:0,1C, 400x9mm, enclosed scale, capillary tube specially coated inside, with non-wetting bl ue special liquid, total immersion, richter-top finish, government calibrated with Calibration Certificate with 5 tes 1 * 1 items
Catalog # AMARN71008-CB/CC
Supplier:  Amarell
Therm. to set Amarell Universal, +48+102:0,1C, 400x9mm, enclosed scale, capillary tube specially coated inside, with non-wetting bl ue special liquid, total immersion, richter-top finish, government calibrated with Calibration Certificate with 5 tes 1 * 1 items
Catalog # AMARN71008-CB/CC
Supplier:  Amarell
Frequently Bought Together