ContactUsLinkComponent
Therm. to set Amarell Universal +48+102:0,1C, 400x9mm, enclosed scale, capillary tube specially coated inside, with non-wetting bl ue special liquid, total immersion, richter-top finish with button, null
Catalog # AMARN71008-CB
Supplier:  Amarell
Therm. to set Amarell Universal +48+102:0,1C, 400x9mm, enclosed scale, capillary tube specially coated inside, with non-wetting bl ue special liquid, total immersion, richter-top finish with button, null
Catalog # AMARN71008-CB
Supplier:  Amarell
Frequently Bought Together