Therm. to set Amarell Universal, -100+30:0,5°C, 400x9mm, enclosed scale, cap.lens front, colourless, toluene, total immersion, richt er-top finish, government verified with Verification Certificate with 3 test points 1 * 1 items
Katalogové číslo AMARN71002-TOL/VC
Supplier: Amarell
Frequently Bought Together