ContactUsLinkComponent
Therm. to set Amarell Universal, -200+30:0,5C, 400x9mm, enclosed scale, cap.lens front, blue reflecting, pentane compound, total im mersion, richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARN71000-PENT
Supplier:  Amarell
Therm. to set Amarell Universal, -200+30:0,5C, 400x9mm, enclosed scale, cap.lens front, blue reflecting, pentane compound, total im mersion, richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARN71000-PENT
Supplier:  Amarell
Frequently Bought Together