ContactUsLinkComponent
Set precision therm., similar ISO 656, short, -38+202:0,2°C, enclosed scale, 12 instruments, in stuffed case, suitable for governmen t verification respectively suitable for government calibration 1 * 1 items
Catalog # AMARN65836-SET
Supplier:  Amarell
Set precision therm., similar ISO 656, short, -38+202:0,2°C, enclosed scale, 12 instruments, in stuffed case, suitable for governmen t verification respectively suitable for government calibration 1 * 1 items
Catalog # AMARN65836-SET
Supplier:  Amarell
Frequently Bought Together