ContactUsLinkComponent
Precision therm., similar ISO 656, short, +190+310:1°C, milk glass scale, cap. colourless, gallium filled, total immersion, max. 250 x9mm, richter-top finish with button, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARN65814-GA
Supplier:  Amarell
Precision therm., similar ISO 656, short, +190+310:1°C, milk glass scale, cap. colourless, gallium filled, total immersion, max. 250 x9mm, richter-top finish with button, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARN65814-GA
Supplier:  Amarell
Frequently Bought Together