ContactUsLinkComponent
Precision therm., similar ISO 656, short, +118+142:0,2°C, milk glass scale, capillary tube specially coated inside, with non-wettin g blue special liquid, total immersion, max. 250x9mm, richter-top finish with button, with ice point scale, null
Catalog # AMARN65756-CB
Supplier:  Amarell
Precision therm., similar ISO 656, short, +118+142:0,2°C, milk glass scale, capillary tube specially coated inside, with non-wettin g blue special liquid, total immersion, max. 250x9mm, richter-top finish with button, with ice point scale, null
Catalog # AMARN65756-CB
Supplier:  Amarell
Frequently Bought Together