ContactUsLinkComponent
Laboratory Hydrometer, DIN 12791 (old), 1,720-1,842:0,001g/cm , ref. temp. 20C, without therm., max. 255mm long, suitable for gover nment verification 1 * 1 items
Catalog # AMARH820314
Supplier:  Amarell
Laboratory Hydrometer, DIN 12791 (old), 1,720-1,842:0,001g/cm , ref. temp. 20C, without therm., max. 255mm long, suitable for gover nment verification 1 * 1 items
Catalog # AMARH820314
Supplier:  Amarell
Frequently Bought Together