ContactUsLinkComponent
Thermometer similar to IP 76 C, solid stem, white backed, 10+55:0,5C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
  AMARA341070-FL
 :  Amarell
Thermometer similar to IP 76 C, solid stem, white backed, 10+55:0,5C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
  AMARA341070-FL
 :  Amarell
Frequently Bought Together