ContactUsLinkComponent
Thermometer similar to IP 60 C, solid stem, white backed, -2+80:0,2°C, capillary tube specially coated inside, with non-wetting blu e special liquid, total immersion, 395x6-8mm, durable pigment, null
Catalog # AMARA340810-CB
Supplier:  Amarell
Thermometer similar to IP 60 C, solid stem, white backed, -2+80:0,2°C, capillary tube specially coated inside, with non-wetting blu e special liquid, total immersion, 395x6-8mm, durable pigment, null
Catalog # AMARA340810-CB
Supplier:  Amarell
Frequently Bought Together