ContactUsLinkComponent
Viscometer-Therm., ähnlich Vogel-Ossag, ähnlich DIN 12785, 0+35:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, 470mm long, lower part solid stem 200x5mm, 208mm immersion, Richter-top finish, null
Catalog # AMARM240102-CB
Supplier:  Amarell
Viscometer-Therm., ähnlich Vogel-Ossag, ähnlich DIN 12785, 0+35:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, 470mm long, lower part solid stem 200x5mm, 208mm immersion, Richter-top finish, null
Catalog # AMARM240102-CB
Supplier:  Amarell
Frequently Bought Together