ContactUsLinkComponent
Viscometer therm., ähnlich Engler, similar DIN, 0+200:1°C, enclosed scale, cap. prismatic, colourless, red special liquid, 300x6mm, 90mm immersion, with swelling, Richter-top finish, without button, suitable for government verification 1 * 1 items
Catalog # AMARM238050-FL
Supplier:  Amarell
Viscometer therm., ähnlich Engler, similar DIN, 0+200:1°C, enclosed scale, cap. prismatic, colourless, red special liquid, 300x6mm, 90mm immersion, with swelling, Richter-top finish, without button, suitable for government verification 1 * 1 items
Catalog # AMARM238050-FL
Supplier:  Amarell
Frequently Bought Together