ContactUsLinkComponent
Flash point therm., similar Marcusson, similar DIN 12785, +40+260:1C, 48mm immersion, 300x9-10mm, milk glass scale, cap. prismatic, colourless, blue special liquid, Richter-top finish, spherical bulb, suitable for government calibration 1 * 1 items
Catalog # AMARM234000-FL
Supplier:  Amarell
Flash point therm., similar Marcusson, similar DIN 12785, +40+260:1C, 48mm immersion, 300x9-10mm, milk glass scale, cap. prismatic, colourless, blue special liquid, Richter-top finish, spherical bulb, suitable for government calibration 1 * 1 items
Catalog # AMARM234000-FL
Supplier:  Amarell
Frequently Bought Together