ContactUsLinkComponent
Flash point-Therm., similar Abel-Pensky, similar DIN, +40+160:0,5°C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. prismat ic, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, suitable for government calibr 1 * 1 items
Catalog # AMARM230026-FL
Supplier:  Amarell
Flash point-Therm., similar Abel-Pensky, similar DIN, +40+160:0,5°C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. prismat ic, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, suitable for government calibr 1 * 1 items
Catalog # AMARM230026-FL
Supplier:  Amarell
Frequently Bought Together