Flash point therm., similar Abel-Pensky, similar DIN 12785, +35+100:1°C, 75mm immersion, max. 285x9-10mm, milk glass scale, cap. pr ismatic, colourless, red special liquid, Richter-top finish, with swelling, cylindrical bulb, null
Catalog # AMARM230002-FL/VC
Supplier: Amarell
Frequently Bought Together