ContactUsLinkComponent
Flash point-Therm., similar Abel-Pensky, similar DIN 12785, 0+70:0,5C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. pris matic, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, suitable for government ver 1 * 1 items
Catalog # AMARM230000-FL
Supplier:  Amarell
Flash point-Therm., similar Abel-Pensky, similar DIN 12785, 0+70:0,5C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. pris matic, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, suitable for government ver 1 * 1 items
Catalog # AMARM230000-FL
Supplier:  Amarell
Frequently Bought Together