ContactUsLinkComponent
Extreme precision therm., with saddle, 0+50:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue sp ecial liquid, 420x9-10mm, total immersion, richter-top finish with button, null
Catalog # AMARL53160-CB
Supplier:  Amarell
Extreme precision therm., with saddle, 0+50:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue sp ecial liquid, 420x9-10mm, total immersion, richter-top finish with button, null
Catalog # AMARL53160-CB
Supplier:  Amarell
Frequently Bought Together