ContactUsLinkComponent
Extreme precision therm., with saddle, -38+50:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, 550x8-9mm, total immersion, richter-top finish with button, null
Catalog # AMARL53154-CB
Supplier:  Amarell
Extreme precision therm., with saddle, -38+50:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, 550x8-9mm, total immersion, richter-top finish with button, null
Catalog # AMARL53154-CB
Supplier:  Amarell
Frequently Bought Together