ContactUsLinkComponent
Extreme precision therm., with saddle, 0+150:0,2°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue s pecial liquid, 480x9-10mm, total immersion, richter-top finish with button, null
Catalog # AMARL53136-CB
Supplier:  Amarell
Extreme precision therm., with saddle, 0+150:0,2°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue s pecial liquid, 480x9-10mm, total immersion, richter-top finish with button, null
Catalog # AMARL53136-CB
Supplier:  Amarell
Frequently Bought Together