ContactUsLinkComponent
Thermometer similar to IP 50 C, solid stem, white backed, 10+65:0,5C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
  AMARA340720-FL
 :  Amarell
Thermometer similar to IP 50 C, solid stem, white backed, 10+65:0,5C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
  AMARA340720-FL
 :  Amarell
Frequently Bought Together