ContactUsLinkComponent
Thermometer similar to IP 50 C, solid stem, white backed, 10+65:0,5C, capillary tube specially coated inside, with non-wetting blu e special liquid, durable pigment, null
Catalog # AMARA340720-CB
Supplier:  Amarell
Thermometer similar to IP 50 C, solid stem, white backed, 10+65:0,5C, capillary tube specially coated inside, with non-wetting blu e special liquid, durable pigment, null
Catalog # AMARA340720-CB
Supplier:  Amarell
Frequently Bought Together