ContactUsLinkComponent
Thermometer similar to IP 49 C, solid stem, white backed, -15+40:0,5C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA340700-FL
Supplier:  Amarell
Thermometer similar to IP 49 C, solid stem, white backed, -15+40:0,5C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA340700-FL
Supplier:  Amarell
Frequently Bought Together