ContactUsLinkComponent
Thermometer similar to IP 46 C, solid stem, white backed, 14,5+21:0,1°C, capillary tube specially coated inside, with non-wetting b lue special liquid, durable pigment, null
Catalog # AMARA340650-CB
Supplier:  Amarell
Thermometer similar to IP 46 C, solid stem, white backed, 14,5+21:0,1°C, capillary tube specially coated inside, with non-wetting b lue special liquid, durable pigment, null
Catalog # AMARA340650-CB
Supplier:  Amarell
Frequently Bought Together