ContactUsLinkComponent
Thermometer similar to IP 44 C, solid stem, white backed, 15+121:0,5°C, capillary tube specially coated inside, with non-wetting bl ue special liquid, durable pigment, null
Catalog # AMARA340620-CB
Supplier:  Amarell
Thermometer similar to IP 44 C, solid stem, white backed, 15+121:0,5°C, capillary tube specially coated inside, with non-wetting bl ue special liquid, durable pigment, null
Catalog # AMARA340620-CB
Supplier:  Amarell
Frequently Bought Together