ContactUsLinkComponent
Thermometer similar to IP 40 C, solid stem, white backed, 20+120:1C, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 100mm, 250x4,5-6mm, null
Catalog # AMARA340570-CB
Supplier:  Amarell
Thermometer similar to IP 40 C, solid stem, white backed, 20+120:1C, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 100mm, 250x4,5-6mm, null
Catalog # AMARA340570-CB
Supplier:  Amarell
Frequently Bought Together