ContactUsLinkComponent
Thermometer similar to IP 38 C, solid stem, white backed, 23+27:0,1C, capillary tube specially coated inside, with non-wetting blu e special liquid, durable pigment, null
Catalog # AMARA340540-CB
Supplier:  Amarell
Thermometer similar to IP 38 C, solid stem, white backed, 23+27:0,1C, capillary tube specially coated inside, with non-wetting blu e special liquid, durable pigment, null
Catalog # AMARA340540-CB
Supplier:  Amarell
Frequently Bought Together