ContactUsLinkComponent
Thermometer similar to IP Abel Water-Bath Celsius, solid stem, white backed, 32+88:0,5°C, capillary tube specially coated inside, w ith non-wetting blue special liquid, sdurable pigment, null
Catalog # AMARA340330-CB
Supplier:  Amarell
Thermometer similar to IP Abel Water-Bath Celsius, solid stem, white backed, 32+88:0,5°C, capillary tube specially coated inside, w ith non-wetting blue special liquid, sdurable pigment, null
Catalog # AMARA340330-CB
Supplier:  Amarell
Frequently Bought Together