ContactUsLinkComponent
Thermometer similar to IP Abel Oil Cup Celsius, solid stem, white backed, 10+65:0,5°C, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA340310-CB
Supplier:  Amarell
Thermometer similar to IP Abel Oil Cup Celsius, solid stem, white backed, 10+65:0,5°C, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA340310-CB
Supplier:  Amarell
Frequently Bought Together