ContactUsLinkComponent
Thermometer similar to IP 24 C, solid stem, white backed, 95+103:0,1°C, capillary tube specially coated inside, with non-wetting bl ue special liquid, total immersion, 275x6-8mm, durable pigment, null
Catalog # AMARA340290-CB
Supplier:  Amarell
Thermometer similar to IP 24 C, solid stem, white backed, 95+103:0,1°C, capillary tube specially coated inside, with non-wetting bl ue special liquid, total immersion, 275x6-8mm, durable pigment, null
Catalog # AMARA340290-CB
Supplier:  Amarell
Frequently Bought Together