ContactUsLinkComponent
Thermometer similar to IP 23 C, solid stem, white backed, 34+42:0,1°C, capillary tube specially coated inside, with non-wetting blu e special liquid, total immersion, 275x6-7mm, durable pigment, null
Catalog # AMARA340270-CB
Supplier:  Amarell
Thermometer similar to IP 23 C, solid stem, white backed, 34+42:0,1°C, capillary tube specially coated inside, with non-wetting blu e special liquid, total immersion, 275x6-7mm, durable pigment, null
Catalog # AMARA340270-CB
Supplier:  Amarell
Frequently Bought Together