ContactUsLinkComponent
Thermometer similar to IP 15 C, solid stem, white backed, -5+110:0,5C, capillary tube specially coated inside, with non-wetting bl ue special liquid, immersion 57mm, max.295x6-7mm, durable pigment, null
Catalog # AMARA340160-CB
Supplier:  Amarell
Thermometer similar to IP 15 C, solid stem, white backed, -5+110:0,5C, capillary tube specially coated inside, with non-wetting bl ue special liquid, immersion 57mm, max.295x6-7mm, durable pigment, null
Catalog # AMARA340160-CB
Supplier:  Amarell
Frequently Bought Together