ContactUsLinkComponent
Setting point therm., DIN 12785, -70+50:1C, milk-glass scale, cap.lens front, colourles, toluene, Richter-top finish without button , upper part 195x10mm, lower part 165x5mm, immersion 180mm, suitable for government verification 1 * 1 items
Catalog # AMARM242020-TOL
Supplier:  Amarell
Setting point therm., DIN 12785, -70+50:1C, milk-glass scale, cap.lens front, colourles, toluene, Richter-top finish without button , upper part 195x10mm, lower part 165x5mm, immersion 180mm, suitable for government verification 1 * 1 items
Catalog # AMARM242020-TOL
Supplier:  Amarell
Frequently Bought Together