Viscometer therm. designed for the ball-draw viscometer, similar DIN 12785, +149+201:0,1°C, enclosed scale, cap. lens front, colourl ess, gallium filled, immersion 140mm, Richter-top finish without button, 460x9,5mm, suitable for government calibrati 1 * 1 items
Catalog # AMARM241042-GA
Supplier: Amarell
Frequently Bought Together