ContactUsLinkComponent
Viscometer therm. designed for the ball-draw viscometer, similar DIN 12785, +49+101:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 140mm, Richter-top finish without button, 460x9,5mm, null
Catalog # AMARM241034-CB
Supplier:  Amarell
Viscometer therm. designed for the ball-draw viscometer, similar DIN 12785, +49+101:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 140mm, Richter-top finish without button, 460x9,5mm, null
Catalog # AMARM241034-CB
Supplier:  Amarell
Frequently Bought Together