ContactUsLinkComponent
Viscometer-Therm., similar Vogel-Ossag, similar DIN, 0+150:1°C, enclosed scale, cap. prismatic, colourless, red special liquid, 470m m long, lower part solid stem 200x5mm, 208mm immersion, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARM240110-FL
Supplier:  Amarell
Viscometer-Therm., similar Vogel-Ossag, similar DIN, 0+150:1°C, enclosed scale, cap. prismatic, colourless, red special liquid, 470m m long, lower part solid stem 200x5mm, 208mm immersion, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARM240110-FL
Supplier:  Amarell
Frequently Bought Together