ContactUsLinkComponent
Well scoop therm., -5+35:0,2C, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, tot al immersion, in slotted brass armour with upper part c.200x20mm and scoop vessel c.70x32mm, null
Catalog # AMARK196120-CB
Supplier:  Amarell
Well scoop therm., -5+35:0,2C, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, tot al immersion, in slotted brass armour with upper part c.200x20mm and scoop vessel c.70x32mm, null
Catalog # AMARK196120-CB
Supplier:  Amarell
Frequently Bought Together