ContactUsLinkComponent
Silo and pit therm., 0+100:1C, special red liquid filled, complete in slotted metal armour with spiked end, upper part of brass 265 x25mm, lower part of steel 300mm, lower part = immersion, with handles 1 * 1 items
Catalog # AMARK190118-FL
Supplier:  Amarell
Silo and pit therm., 0+100:1C, special red liquid filled, complete in slotted metal armour with spiked end, upper part of brass 265 x25mm, lower part of steel 300mm, lower part = immersion, with handles 1 * 1 items
Catalog # AMARK190118-FL
Supplier:  Amarell
Frequently Bought Together