ContactUsLinkComponent
Silo and pit therm., -38+50:1C, special red liquid filled, complete in slotted metal armour with spiked end, upper part of brass 26 5x25mm, lower part of steel 300mm, lower part = immersion, with handles 1 * 1 items
  AMARK190100-FL
 :  Amarell
Silo and pit therm., -38+50:1C, special red liquid filled, complete in slotted metal armour with spiked end, upper part of brass 26 5x25mm, lower part of steel 300mm, lower part = immersion, with handles 1 * 1 items
  AMARK190100-FL
 :  Amarell
Frequently Bought Together