ContactUsLinkComponent
Soil therm., simple type, -5+83:1C, solid stem, white backed, red filling, immersion 75mm, in green plastic case with red button, c .320mm long 1 * 1 items
Catalog # AMARK189500-FL
Supplier:  Amarell
Soil therm., simple type, -5+83:1C, solid stem, white backed, red filling, immersion 75mm, in green plastic case with red button, c .320mm long 1 * 1 items
Catalog # AMARK189500-FL
Supplier:  Amarell
Frequently Bought Together