ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 122 C, white backed, -45-35:0,1C, capillary tube specially coated inside, with non-wettin g blue special liquid, total immersion, max.305x5,5-8mm, null
Catalog # AMARA301650-CB
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 122 C, white backed, -45-35:0,1C, capillary tube specially coated inside, with non-wettin g blue special liquid, total immersion, max.305x5,5-8mm, null
Catalog # AMARA301650-CB
Supplier:  Amarell
Frequently Bought Together