ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 121 C, white backed, 98,6+101,4:0,05C, capillary tube specially coated inside, with non-w etting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA301630-CB
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 121 C, white backed, 98,6+101,4:0,05C, capillary tube specially coated inside, with non-w etting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA301630-CB
Supplier:  Amarell
Frequently Bought Together