ContactUsLinkComponent
Precision therm., straight stem, similar DIN, -20+360:2C, cap. lens front, colourless, Eco-friendly, green liquid, biodegradable, u pper part 275x11mm, lower part 100x8mm, suitable for government calibration 1 * 1 items
Catalog # AMARL36772-EF
Supplier:  Amarell
Precision therm., straight stem, similar DIN, -20+360:2C, cap. lens front, colourless, Eco-friendly, green liquid, biodegradable, u pper part 275x11mm, lower part 100x8mm, suitable for government calibration 1 * 1 items
Catalog # AMARL36772-EF
Supplier:  Amarell
Frequently Bought Together