ContactUsLinkComponent
Precision therm., straight stem, similar DIN, -20+250:1C, cap. prismatic, colourless, blue special liquid, upper part 275x11mm, low er part 300x8mm, suitable for government calibration 1 * 1 items
Catalog # AMARL36760-FL
Supplier:  Amarell
Precision therm., straight stem, similar DIN, -20+250:1C, cap. prismatic, colourless, blue special liquid, upper part 275x11mm, low er part 300x8mm, suitable for government calibration 1 * 1 items
Catalog # AMARL36760-FL
Supplier:  Amarell
Frequently Bought Together