ContactUsLinkComponent
Precision therm., straight stem, 0+100:0,1C, capillary tube specially coated inside, with non-wetting blue special liquid, upper p art 520x11mm, lower part 100x7-8mm, null
Catalog # AMARL36070-CB
Supplier:  Amarell
Precision therm., straight stem, 0+100:0,1C, capillary tube specially coated inside, with non-wetting blue special liquid, upper p art 520x11mm, lower part 100x7-8mm, null
Catalog # AMARL36070-CB
Supplier:  Amarell
Frequently Bought Together