ContactUsLinkComponent
Precision therm., straight stem, -10+50:0,1C, capillary tube specially coated inside, with non-wetting blue special liquid, upper part 350x11mm, lower part 100x7-8mm, null
Catalog # AMARL36040-CB
Supplier:  Amarell
Precision therm., straight stem, -10+50:0,1C, capillary tube specially coated inside, with non-wetting blue special liquid, upper part 350x11mm, lower part 100x7-8mm, null
Catalog # AMARL36040-CB
Supplier:  Amarell
Frequently Bought Together